(S)-tert-Butyl 3-(hydroxymethyl)piperazine-1-carboxylate,98% 1 * 5 g
AMBDA109002-5G
: AMBEED
Frequently Bought Together
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
: VWR Chemicals