4-(t-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid,98% 1 * 0,25 g
AMBDA507055-250MG
: AMBEED
4-(t-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid,98% 1 * 0,25 g
AMBDA507055-250MG
: AMBEED
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.0% Ph. Eur.
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals