4-Methoxy-N-methylbenzylamine,98% 1 * 10 g
Catalog # AMBDA326303-10G
Supplier: AMBEED
Frequently Bought Together
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-Dichloroethane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.7% for gas chromatography
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
Supplier: Honeywell Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific