Hexyl methanesulfonate,97% 1 * 5 g
Catalog # AMBDA129136-5G
Supplier: AMBEED
Frequently Bought Together
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
4-(Methylamino)phenol sulphate 99.0-101.5% ACS
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Trifluoroacetic Acid for UV 1 * 100 mL
Supplier: PanReac AppliChem
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Isobutanol, Sigma-Aldrich®
Supplier: Merck