Methyl methylcarbamate,95% 1 * 5 g
AMBDA132655-5G
: AMBEED
Frequently Bought Together
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
n-Hexane 95+%, extra pure
: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Dichloromethane, anhydrous ≥99.7% stabilised
: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
: Thermo Fisher Scientific
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+%, extra pure
: Thermo Fisher Scientific
n-Heptane, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
: Honeywell Chemicals
Acetone ≥99%
: Thermo Fisher Scientific