tert-Butyl 3-bromo-4-oxopiperidine-1-carboxylate,95% 1 * 25 g
Catalog # AMBDA207023-25G
Supplier: AMBEED
tert-Butyl 3-bromo-4-oxopiperidine-1-carboxylate,95% 1 * 25 g
Catalog # AMBDA207023-25G
Supplier: AMBEED
Frequently Bought Together
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck