Boc-D-Phenylglycinol,97% 1 * 10 g
AMBDA227101-10G
: AMBEED
Frequently Bought Together
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Diethylene glycol for synthesis
: Merck
m-Xylene, extra dry 99%, AcroSeal®
: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
: Thermo Fisher Scientific
Toluene, extra dry 99.85%, AcroSeal®
: Thermo Fisher Scientific
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
: Thermo Fisher Scientific
1-Propanol, anhydrous ≥99.9%
: Thermo Fisher Scientific
1-Butanol ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Dimethyl sulphoxide ≥99.5%, Reagent Grade
: Honeywell Chemicals
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
: Honeywell Chemicals
1,4-Dioxane ≥99% stabilised
: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
: Honeywell Chemicals
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
: Thermo Fisher Scientific