1-Cbz-3-Hydroxyazetidine,97% 1 * 25 g
AMBDA162071-25G
: AMBEED
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals