6-Bromopicolinonitrile,98% 1 * 5 g
Catalog # AMBDA339033-5G
Supplier: AMBEED
Frequently Bought Together
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Supplier: Merck
Isobutanol, Sigma-Aldrich®
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
n-Hexane
Supplier: Roth Carl
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.5%, extra pure
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals