(4-(tert-Butylthio)phenyl)boronic acid,98% 1 * 0,1 g
Artikelnr. AMBDA406046-100MG
Supplier: AMBEED
Frequently Bought Together
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Iodomethane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Benzoic acid methyl ester, EMPLURA®, Supelco®
Supplier: Merck
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals