(4-Bromo-2,3,5,6-tetrafluorophenyl)boronic acid,95% 1 * 0,1 g
Catalog # AMBDA180847-100MG
Supplier: AMBEED
(4-Bromo-2,3,5,6-tetrafluorophenyl)boronic acid,95% 1 * 0,1 g
Catalog # AMBDA180847-100MG
Supplier: AMBEED
Frequently Bought Together
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
(±)-1,2-Propanediol ≥99.5%, meets analytical specification of BP, Ph. Eur., USP, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals