(3-Bromo-6-chloropyridin-2-yl)methanol,97% 1 * 0,1 g
Catalog # AMBDA119250-100MG
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Turpentine oil
Supplier: Roth Carl
Dimethyl Sulfoxide, Reagent ACS Grade 1 * 500 mL
Supplier: Spectrum Chemical
1-Chlorobutane ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile
Supplier: Roth Carl
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Butanone (Methylethylketone) pure 1 * 2,5 L
Supplier: PanReac AppliChem
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
n-Hexane
Supplier: Roth Carl