(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxyphenyl)propanoate,95% 1 * 10 g
Catalog # AMBDA175473-10G
Supplier: AMBEED
(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxyphenyl)propanoate,95% 1 * 10 g
Catalog # AMBDA175473-10G
Supplier: AMBEED
Frequently Bought Together
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
Supplier: PanReac AppliChem
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran stabilised ACS, Burdick & Jackson™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
Acetone ≥99.5% for spectrophotometry
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals