2-Bromo-5-fluoroisonicotinonitrile,95% 1 * 0,25 g
Catalog # AMBDA267155-250MG
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Supplier: Honeywell Chemicals
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
PETROLEUM ETHER 40-60°C 1 * 5 L
Supplier: Roth Carl
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethylenediamine, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Aeroflow tap, na, general spare part 1 * 1 items
Supplier: Avantor
Orthophosphoric acid 10%
Supplier: Hach