4,5-Dibromo-2-methyl-2H-1,2,3-triazole,97% 1 * 0,1 g
Catalog # AMBDA172490-100MG
Supplier: AMBEED
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Benzoic acid methyl ester
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
GLYCEROL PURISS., MEETS ANALYTICAL SPECI 1 * 1 L
Supplier: Merck
Glycerine, Hi-LR™
Supplier: HIMEDIA
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific