1,4-Dimethyl-1H-pyrazol-3-amine,98% 1 * 1 g
Catalog # AMBDA106669-1G
Supplier: AMBEED
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals