1-(4-Bromo-2-fluorophenyl)propan-1-one,95% 1 * 0,1 g
Catalog # AMBDA153511-100MG
Supplier: AMBEED
Frequently Bought Together
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
1,2,3,4-TETRAHYDRONAPHTHALENE FOR SYN... 1 * 200 kg
Supplier: Merck
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethyl acetate, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Trichloroethylene 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
Iodomethane stabilised, Sigma-Aldrich®
Supplier: Merck
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
Water for LC-MS 1 * 2,5 L
Supplier: PanReac AppliChem
n-Heptane 99+%, pure
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific