N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide,97% 1 * 0,25 g
Catalog # AMBDA152370-250MG
Supplier: AMBEED
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide,97% 1 * 0,25 g
Catalog # AMBDA152370-250MG
Supplier: AMBEED
Frequently Bought Together
Benzoic acid methyl ester, EMPLURA®, Supelco®
Supplier: Merck
Acetone ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND