tert-Butyl 6-bromo-1H-indole-1-carboxylate,96% 1 * 0,25 g
Catalog # AMBDA510765-250MG
Supplier: AMBEED
tert-Butyl 6-bromo-1H-indole-1-carboxylate,96% 1 * 0,25 g
Catalog # AMBDA510765-250MG
Supplier: AMBEED
Frequently Bought Together
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dichloromethane 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.3%, Laboratory Reagent
Supplier: Honeywell Chemicals
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals