6-Methyl-2-vinyl-1,3,6,2-dioxazaborocane-4,8-dione,95% 1 * 0,25 g
Catalog # AMBDA147233-250MG
Supplier: AMBEED
6-Methyl-2-vinyl-1,3,6,2-dioxazaborocane-4,8-dione,95% 1 * 0,25 g
Catalog # AMBDA147233-250MG
Supplier: AMBEED
Frequently Bought Together
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
(±)-1,2-Propanediol ACS
Supplier: Thermo Fisher Scientific
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific