(R)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid,98% 1 * 0,1 g
Catalog # AMBDA681574-100MG
Supplier: AMBEED
(R)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid,98% 1 * 0,1 g
Catalog # AMBDA681574-100MG
Supplier: AMBEED
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific