3-(Hydroxymethyl)-5-methylphenylboronic Acid,98% 1 * 0,1 g
Catalog # AMBDA190047-100MG
Supplier: AMBEED
3-(Hydroxymethyl)-5-methylphenylboronic Acid,98% 1 * 0,1 g
Catalog # AMBDA190047-100MG
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Tetrachloroethylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Chloroform, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Dibutyl ether anhydrous, 99.3 % 1 * 2 L
Supplier: Merck
Petroleum spirit, 100…120 °C, AnalaR NORMAPUR® ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
9-Fluorenylmethyl chloroformate (Fmoc chloride), Supelco®
Supplier: Merck
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Chloroform ≥99.5% for spectrophotometry
Supplier: Thermo Fisher Scientific