(6-Ethoxy-5-methylpyridin-3-yl)boronic acid,98% 1 * 0,25 g
Catalog # AMBDA887371-250MG
Supplier: AMBEED
(6-Ethoxy-5-methylpyridin-3-yl)boronic acid,98% 1 * 0,25 g
Catalog # AMBDA887371-250MG
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
p-Xylene 99% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals