(2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid),98% 1 * 100 g
Catalog # AMBDA160767-100G
Supplier: AMBEED
(2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid),98% 1 * 100 g
Catalog # AMBDA160767-100G
Supplier: AMBEED
Frequently Bought Together
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
n-Heptane, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Propanol ≥99.7% for gas chromatography
Supplier: Thermo Fisher Scientific
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Formamide ≥99%
Supplier: Thermo Fisher Scientific