2-Hydroxy-5-octanoylbenzoic acid,97% 1 * 25 g
Catalog # AMBDA132116-25G
Supplier: AMBEED
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetonitrile
Supplier: Roth Carl