3-Phenylacrylaldehyde,98% 1 * 25 g
Catalog # AMBDA331599-25G
Supplier: AMBEED
Frequently Bought Together
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Tetrachloroethylene, EMPLURA®, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Supplier: VWR Chemicals
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Butyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
N,N-Diethyl-p-phenylenediammonium sulphate, Supelco®
Supplier: Merck
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol), extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific