(1H-Pyrazol-3-yl)methanamine,97% 1 * 0,25 g
Catalog # AMBDA136013-250MG
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Safety Bottle Container
Supplier: Merck
Cyclohexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥99%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor