(4-Morpholinophenyl)boronic acid,98% 1 * 0,25 g
Catalog # AMBDA418226-250MG
Supplier: AMBEED
Frequently Bought Together
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Isopropyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Chlorobenzene, Sigma-Aldrich®
Supplier: Merck
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific