3-(p-Tolyl)propionic Acid,98% 1 * 5 g
AMBDA879552-5G
: AMBEED
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
: VWR Chemicals
: VWR Chemicals
: VWR Chemicals