ETHYL 4-BROMO-3-OXOBUTANOATE 1 * 5 g
AMBDA556966-5G
: AMBEED
Frequently Bought Together
Aceton ≥99,5%, GPR RECTAPUR®
: VWR Chemicals
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Wasser, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, rein
: VWR Chemicals
Xylol (Isomerengemisch) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Aceton ≥99,8%, HiPerSolv CHROMANORM® für die HPLC
: VWR Chemicals