4-Fluoro-3-methoxybenzaldehyde,98% 1 * 25 g
Catalog # AMBDA476397-25G
Supplier: AMBEED
Frequently Bought Together
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-DIMETHOXYETHANE 1 * 1 L
Supplier: Merck
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals