4-(Dimethylamino)benzaldehyde,97% 1 * 100 g
Catalog # AMBDA128847-100G
Supplier: AMBEED
Frequently Bought Together
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Butyl acetate 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.8% ACS
Supplier: Thermo Fisher Scientific
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Cyclohexane for HPLC
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Liquid for manometer with sloping tube
Supplier: VWR Chemicals