1-Cyclopropylpiperazine,97% 1 * 5 g
Catalog # AMBDA187680-5G
Supplier: AMBEED
Frequently Bought Together
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide, EMPLURA®, Supelco®
Supplier: Merck
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, anhydrous (max. 0.005% H₂O)
Supplier: VWR Chemicals
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
N-Methyl-2-pyrrolidone (NMP), extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclopentane 95+%, pure
Supplier: Thermo Fisher Scientific
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals