2-Bromo-1-chloro-3-nitrobenzene,98% 1 * 5 g
Catalog # AMBDA135451-5G
Supplier: AMBEED
Frequently Bought Together
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.5% (by GC), EMPROVE® ESSENTIAL NF, Ph. Eur., BP, JPE, extra pure suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Diethyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol denatured ≥96%, extra pure (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-ETHOXYETHANOL, REAGENTPLUS, 99%. 1 * 1 Each
Supplier: Merck
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals