Methyl 5-bromopyrimidine-2-carboxylate,95% 1 * 10 g
AMBDA281111-10G
: AMBEED
Frequently Bought Together
Methanol, SupraSolv® for GC-MS
: Merck
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals