tert-Butyl 4-bromo-2-nitrobenzoate,97% 1 * 0,25 g
Artikelnr. AMBDA293145-250MG
Lieferant: AMBEED
Frequently Bought Together
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
Water, HiPerSolv CHROMANORM®, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Lieferant: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Lieferant: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Methyl ethyl ketone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
Lieferant: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Lieferant: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Lieferant: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Lieferant: VWR Chemicals