tert-Butyl trichloroacetimidate,98% 1 * 10 g
Catalog # AMBDA817163-10G
Supplier: AMBEED
Frequently Bought Together
Dichloromethane stabilised Ph. Eur.
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
1,3,5-Trioxane for synthesis, Sigma-Aldrich®
Supplier: Merck
Petroleum spirit, 40…60 °C, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
Methanol, LiChrosolv® tested for UHPLC-MS, Supelco®
Supplier: Merck
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific