Methyl 2-chloropyrimidine-5-carboxylate,98% 1 * 5 g
Catalog # AMBDA177135-5G
Supplier: AMBEED
Frequently Bought Together
n-Pentane ≥99.5% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor