Methyl2-(4-isobutylphenyl)propanoate,97% 1 * 25 g
AMBDA508535-25G
: AMBEED
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, TECHNICAL, demineralised
: VWR Chemicals
Acetonitrile ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Tetrahydrofuran, anhydrous 99.9% stabilised, extra pure
: Thermo Fisher Scientific
n-Hexane ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Water, extra pure, Deionized
: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Isobutanol 99+% ACS
: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Acetonitrile, extra dry 99.9+%, AcroSeal®
: Thermo Fisher Scientific
n-Hexane, extra dry 96+%, AcroSeal®
: Thermo Fisher Scientific