tert-Butyl 4-(2-aminoethyl)piperazine-1-carboxylate,97% 1 * 10 g
Catalog # AMBDA113040-10G
Supplier: AMBEED
tert-Butyl 4-(2-aminoethyl)piperazine-1-carboxylate,97% 1 * 10 g
Catalog # AMBDA113040-10G
Supplier: AMBEED
Frequently Bought Together
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Acetonitrile ≥99.7%, Environmental Grade
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Decane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Acetonitrile, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Butyl acetate ≥99.5%, Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor