1,3-Dicyanobenzene,98% 1 * 25 g
AMBDA310400-25G
: AMBEED
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Glycerine 85% Ph. Eur.
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals