1-(Cyclohexylmethyl)piperazine,95% 1 * 5 g
Catalog # AMBDA583162-5G
Supplier: AMBEED
Frequently Bought Together
Sulphur trioxide pyridine complex for synthesis
Supplier: Merck
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane
Supplier: Roth Carl
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Iodomethane stabilised, Sigma-Aldrich®
Supplier: Merck
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals