Hydrazine hydrochloride,97% 1 * 25 g
Catalog # AMBDA681687-25G
Supplier: AMBEED
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Supplier: Merck
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetone, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabiliser free, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals