1-Ethynyl-4-propylbenzene,97% 1 * 25 g
AMBDA209453-25G
: AMBEED
Frequently Bought Together
Benzyl alcohol ≥99.0% analytical reagent
: VWR Chemicals
n-Heptane, EMPLURA®, Supelco®
: Merck
n-Hexane
: Roth Carl
Toluene 99.8+% for analysis
: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
: Honeywell Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
: Thermo Fisher Scientific
Cyclohexane 99+% for spectroscopy
: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
: Thermo Fisher Scientific
n-Hexane
: Roth Carl
Acetonitrile Reag. Ph. Eur., gradient grade for HPLC, Burdick & Jackson™
: Honeywell Chemicals