Ethyl 2-(tetrahydro-2H-pyran-4-yl)acetate,98% 1 * 0,1 g
Catalog # AMBDA119655-100MG
Supplier: AMBEED
Ethyl 2-(tetrahydro-2H-pyran-4-yl)acetate,98% 1 * 0,1 g
Catalog # AMBDA119655-100MG
Supplier: AMBEED
Frequently Bought Together
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
2-Methylbutane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
n-Hexane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
Benzonitrile for HPLC
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran
Supplier: Merck