Ethyl 4-aminobutanoate hydrochloride,98% 1 * 5 g
Catalog # AMBDA121091-5G
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane
Supplier: Roth Carl
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N-HEPTANE 1 * 1 L
Supplier: Roth Carl
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem