Sodium 3-hydroxybutanoate,98% 1 * 5 g
Catalog # AMBDA806846-5G
Supplier: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99%
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
p-Xylene 99%, pure
Supplier: Thermo Fisher Scientific
Glycerine, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Acetonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific