4-Cyclohexylbutan-1-ol,95% 1 * 5 g
Catalog # AMBDA490697-5G
Supplier: AMBEED
Frequently Bought Together
1-Octanol, EMPLURA®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Hexane, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Cyclohexane, EMPLURA®, Supelco®
Supplier: Merck
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem