(S)-4-Phenyloxazolidin-2-one,95% 1 * 10 g
Catalog # AMBDA462404-10G
Supplier: AMBEED
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Supplier: Merck
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Ethylene glycol 99.5% for analysis
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Water, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
Ethanolamine, Sigma-Aldrich®
Supplier: Merck