1-Fluoro-4-(2-bromoethyl)benzene,98% 1 * 5 g
Catalog # AMBDA152452-5G
Supplier: AMBEED
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane
Supplier: Roth Carl
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2-Ethoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
n-Hexane
Supplier: Roth Carl
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals